Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Itm2c
Entrez Gene ID
301575
Gene Name
integral membrane protein 2C
Gene Aliases
-
Chromosome Location
Chr.9: 92916469 - 92930306 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009674.1 NP_001009674.1 5 - 6 789 87
GenBank mRNA BC087127.1 - 5 - 6 789 87

View Full Product Details