Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zpbp
Entrez Gene ID
498415
Gene Name
zona pellucida binding protein
Gene Aliases
-
Chromosome Location
Chr.14: 91359751 - 91556306 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025139.1 NP_001020310.1 6 - 7 1029 76
XM_006251505.2 XP_006251567.1 6 - 7 1059 76
GenBank mRNA BC079315.1 - 6 - 7 1029 76

View Full Product Details