The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012235.1 | NP_001012235.1 | 10 - 11 | 977 | 92 |
GenBank mRNA | AY574213.1 | - | 10 - 11 | 977 | 92 |