The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108471.1 | NP_001101941.1 | 5 - 6 | 981 | 91 |
GenBank mRNA | FQ209516.1 | - | 6 - 7 | 574 | 91 |