Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chmp2b
Entrez Gene ID
363720
Gene Name
charged multivesicular body protein 2B
Gene Aliases
RGD1306781
Chromosome Location
Chr.11: 2665264 - 2692254 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_343995.7 XP_343996.5 3 - 4 616 141
GenBank mRNA FQ212834.1 - 3 - 4 581 141

View Full Product Details