The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108886.1 | NP_001102356.1 | 3 - 4 | 400 | 66 |
GenBank mRNA | FQ228021.1 | - | 3 - 4 | 371 | 66 |
FQ233353.1 | - | 3 - 4 | 367 | 66 |