The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019142.2 | NP_062015.2 | 8 - 9 | 1427 | 127 |
GenBank mRNA | U40819.1 | - | 8 - 9 | 1394 | 127 |