Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tma16
Entrez Gene ID
290686
Gene Name
translation machinery associated 16 homolog (S. cerevisiae)
Gene Aliases
RGD1305222
Chromosome Location
Chr.16: 24978431 - 24997286 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006253023.2 XP_006253085.1 1 - 2 143 123

View Full Product Details