Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Limd1
Entrez Gene ID
316101
Gene Name
LIM domains containing 1
Gene Aliases
-
Chromosome Location
Chr.8: 132553218 - 132598826 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112737.2 NP_001106208.1 4 - 5 1997 82
GenBank mRNA BC168666.1 - 4 - 5 1997 82

View Full Product Details