Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rfc4
Entrez Gene ID
288003
Gene Name
replication factor C (activator 1) 4
Gene Aliases
-
Chromosome Location
Chr.11: 81358592 - 81373044 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105869.2 NP_001099339.1 5 - 6 440 152
GenBank mRNA BC168166.1 - 5 - 6 440 152
FQ228232.1 - 5 - 6 472 152

View Full Product Details