Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl22
Entrez Gene ID
303792
Gene Name
kelch-like family member 22
Gene Aliases
RGD1306288
Chromosome Location
Chr.11: 87646484 - 87718808 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107079.1 NP_001100549.1 5 - 6 1373 76
XM_006248699.2 XP_006248761.1 5 - 6 1715 76
XM_006248700.2 XP_006248762.1 5 - 6 1715 76
XM_006248701.2 XP_006248763.1 6 - 7 1602 76

View Full Product Details