Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dgcr14
Entrez Gene ID
360741
Gene Name
DiGeorge syndrome critical region gene 14
Gene Aliases
-
Chromosome Location
Chr.11: 87224373 - 87234329 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012472.1 NP_001012490.1 3 - 4 416 97
XM_006248636.2 XP_006248698.1 3 - 4 464 97
GenBank mRNA BC089898.1 - 3 - 4 416 97

View Full Product Details