Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gsc2
Entrez Gene ID
363831
Gene Name
goosecoid homeobox 2
Gene Aliases
Gscl
Chromosome Location
Chr.11: 87220618 - 87222606 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108846.1 NP_001102316.1 1 - 2 285 97

View Full Product Details