Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Parl
Entrez Gene ID
287979
Gene Name
presenilin associated, rhomboid-like
Gene Aliases
Psarl
Chromosome Location
Chr.11: 84517368 - 84544463 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035249.1 NP_001030326.1 1 - 2 156 98
GenBank mRNA BC088226.1 - 1 - 2 113 98
BC105907.1 - 1 - 2 156 98
BC126098.1 - 1 - 2 132 98
BC158564.1 - 1 - 2 140 98
FQ214115.1 - 1 - 2 151 98

View Full Product Details