Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcdhga1
Entrez Gene ID
553129
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
-
Chromosome Location
Chr.18: 30808404 - 30971113 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037140.1 NP_001032217.1 - 2425 76
GenBank mRNA AY574008.1 - - 2425 76

View Full Product Details