The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012945.1 | NP_037077.1 | 2 - 3 | 245 | 63 |
GenBank mRNA | L05489.1 | - | 2 - 3 | 245 | 63 |