Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf14
Entrez Gene ID
619577
Gene Name
ring finger protein 14
Gene Aliases
ARA54
Chromosome Location
Chr.18: 31443432 - 31468641 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034995.1 NP_001030167.1 3 - 4 396 110
XM_006254636.2 XP_006254698.1 3 - 4 490 110
XM_006254637.2 XP_006254699.1 3 - 4 804 110
XM_006254639.2 XP_006254701.1 3 - 4 492 110
XM_008772069.1 XP_008770291.1 3 - 4 356 110
GenBank mRNA BC103648.1 - 3 - 4 396 110

View Full Product Details