Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf14
Entrez Gene ID
619577
Gene Name
ring finger protein 14
Gene Aliases
ARA54
Chromosome Location
Chr.18: 31443432 - 31468641 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006254640.2 XP_006254702.1 3 - 4 488 105

View Full Product Details