Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syt4
Entrez Gene ID
64440
Gene Name
synaptotagmin IV
Gene Aliases
-
Chromosome Location
Chr.18: 24172580 - 24181989 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031693.2 NP_113881.1 2 - 3 1081 100
GenBank mRNA DQ181551.1 - 2 - 3 845 100
FQ211592.1 - 2 - 3 1078 100
L38247.1 - 2 - 3 1082 100
U14398.1 - 2 - 3 1110 100

View Full Product Details