Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Matr3
Entrez Gene ID
29150
Gene Name
matrin 3
Gene Aliases
P130/MAT3
Chromosome Location
Chr.18: 28361228 - 28390764 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019149.3 NP_062022.2 4 - 5 1256 69
XM_006254534.2 XP_006254596.1 4 - 5 1278 69
XM_006254535.2 XP_006254597.1 4 - 5 1229 69
GenBank mRNA AB205483.1 - 3 - 4 2026 69
BC062231.1 - 4 - 5 1279 69
BC168723.1 - 4 - 5 1228 69
M63485.1 - 4 - 5 1241 69

View Full Product Details