Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pkd2l2
Entrez Gene ID
291683
Gene Name
polycystic kidney disease 2-like 2
Gene Aliases
-
Chromosome Location
Chr.18: 27190661 - 27225488 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106156.1 NP_001099626.1 1 - 2 78 61
XM_008772026.1 XP_008770248.1 1 - 2 391 61
XM_008772027.1 XP_008770249.1 1 - 2 391 61

View Full Product Details