Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RGD735029
Entrez Gene ID
307480
Gene Name
SEL1 domain containing protein RGD735029
Gene Aliases
Dele
Chromosome Location
Chr.18: 31396898 - 31409914 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199493.1 NP_955787.1 6 - 7 704 113
XM_006254634.2 XP_006254696.1 6 - 7 811 113
GenBank mRNA BC064660.1 - 6 - 7 704 113
FQ211991.1 - - 760 113

View Full Product Details