Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hars2
Entrez Gene ID
307491
Gene Name
histidyl-tRNA synthetase 2, mitochondrial
Gene Aliases
Hars2l, RGD1308426, Zmat2
Chromosome Location
Chr.18: 29629201 - 29638460 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014012.2 NP_001014034.2 8 - 9 942 70
XM_006254575.2 XP_006254637.1 7 - 8 852 70
GenBank mRNA BC089973.1 - 1 - 2 106 70
CV075096.1 - 5 - 6 545 70

View Full Product Details