Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr55
Entrez Gene ID
307494
Gene Name
WD repeat domain 55
Gene Aliases
LRRG00133, RGD1305640
Chromosome Location
Chr.18: 29605217 - 29608730 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017932.1 NP_001017932.1 6 - 7 917 56
GenBank mRNA BC127460.1 - 6 - 7 872 56

View Full Product Details