Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hmgn1
Entrez Gene ID
360704
Gene Name
high mobility group nucleosome binding domain 1
Gene Aliases
-
Chromosome Location
Chr.11: 36474028 - 36479947 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013184.1 NP_001013202.1 1 - 2 99 48
GenBank mRNA BC086423.1 - 1 - 2 99 48
FQ220876.1 - 1 - 2 137 48
FQ222121.1 - 1 - 2 138 48
FQ230156.1 - 1 - 2 102 48

Gene Symbol
LOC100911295
Entrez Gene ID
100911295
Gene Name
non-histone chromosomal protein HMG-14-like
Gene Aliases
-
Chromosome Location
Chr.11: 39482337 - 39488327 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751029.3 XP_003751077.1 1 - 2 170 48

View Full Product Details