The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017177.1 | NP_058873.1 | 10 - 11 | 1387 | 68 |
XM_006242181.2 | XP_006242243.2 | 10 - 11 | 2053 | 68 | |
XM_008765677.1 | XP_008763899.1 | 10 - 11 | 2050 | 68 | |
GenBank mRNA | AB006607.1 | - | 10 - 11 | 1387 | 68 |
BC060515.1 | - | 10 - 11 | 1363 | 68 |