The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108732.1 | NP_001102202.1 | 2 - 3 | 126 | 92 |
XM_006240958.2 | XP_006241020.1 | 2 - 3 | 138 | 92 | |
XM_008765118.1 | XP_008763340.1 | 2 - 3 | 128 | 92 |