Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mbd3
Entrez Gene ID
362834
Gene Name
methyl-CpG binding domain protein 3
Gene Aliases
-
Chromosome Location
Chr.7: 12179046 - 12186251 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108735.1 NP_001102205.1 3 - 4 710 75
XM_006240965.1 XP_006241027.1 3 - 4 704 75
XM_006240966.1 XP_006241028.1 3 - 4 603 75
XM_008765119.1 XP_008763341.1 4 - 5 582 75
XM_008765120.1 XP_008763342.1 3 - 4 522 75
XM_008765121.1 XP_008763343.1 4 - 5 506 75
XM_008765122.1 XP_008763344.1 3 - 4 702 75
GenBank mRNA BC167041.1 - 3 - 4 477 75
FQ216718.1 - 2 - 3 218 75

View Full Product Details