Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddx18
Entrez Gene ID
308490
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 18
Gene Aliases
-
Chromosome Location
Chr.13: 37522558 - 37536066 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006996.1 NP_001006997.1 4 - 5 696 84
GenBank mRNA BC083919.1 - 4 - 5 696 84

View Full Product Details