The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006996.1 | NP_001006997.1 | 10 - 11 | 1683 | 71 |
GenBank mRNA | BC083919.1 | - | 10 - 11 | 1683 | 71 |