Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ribc1
Entrez Gene ID
317431
Gene Name
RIB43A domain with coiled-coils 1
Gene Aliases
-
Chromosome Location
Chr.X: 21699361 - 21711345 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006256801.2 XP_006256863.1 1 - 2 224 78

View Full Product Details