Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B3galtl
Entrez Gene ID
689765
Gene Name
beta 1,3-galactosyltransferase-like
Gene Aliases
B3galtl-ps1
Chromosome Location
Chr.12: 6385830 - 6476010 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008769054.1 XP_008767276.1 6 - 7 922 91

View Full Product Details