Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gabrg3
Entrez Gene ID
79211
Gene Name
gamma-aminobutyric acid (GABA) A receptor, gamma 3
Gene Aliases
-
Chromosome Location
Chr.1: 112158530 - 112811954 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024370.3 NP_077346.3 2 - 3 321 149
GenBank mRNA M81142.1 - 2 - 3 303 149
X63324.1 - - 377 149

View Full Product Details