Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plekha8
Entrez Gene ID
500132
Gene Name
pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 8
Gene Aliases
FAPP-2, RGD1563940
Chromosome Location
Chr.4: 84770290 - 84820887 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109235.2 NP_001102705.1 6 - 7 804 98
XM_006236506.2 XP_006236568.1 6 - 7 889 98

View Full Product Details