Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppm1k
Entrez Gene ID
312381
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent, 1K
Gene Aliases
RGD1308501
Chromosome Location
Chr.4: 88694370 - 88721374 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107863.1 NP_001101333.1 1 - 2 201 124

View Full Product Details