The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126296.2 | NP_001119768.2 | 6 - 7 | 927 | 87 |
GenBank mRNA | BC161978.1 | - | 6 - 7 | 927 | 87 |
FQ226359.1 | - | 1 - 2 | 83 | 87 |