Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrrc61
Entrez Gene ID
500111
Gene Name
leucine rich repeat containing 61
Gene Aliases
RGD1561800
Chromosome Location
Chr.4: 78185946 - 78206818 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109231.1 NP_001102701.1 3 - 4 1225 78

View Full Product Details