Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nom1
Entrez Gene ID
682058
Gene Name
nucleolar protein with MIF4G domain 1
Gene Aliases
-
Chromosome Location
Chr.4: 2340034 - 2356427 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008762677.1 XP_008760899.1 1 - 2 989 110

View Full Product Details