Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tectb
Entrez Gene ID
292124
Gene Name
tectorin beta
Gene Aliases
-
Chromosome Location
Chr.1: 275957698 - 275976572 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106200.1 NP_001099670.1 6 - 7 795 70

View Full Product Details