Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mxi1
Entrez Gene ID
25701
Gene Name
MAX interactor 1
Gene Aliases
MXI-WR
Chromosome Location
Chr.1: 274030748 - 274090071 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013160.2 NP_037292.2 5 - 6 564 90
XM_006231604.1 XP_006231666.1 5 - 6 1123 90
XM_006231605.1 XP_006231667.1 5 - 6 535 90
XM_006231606.1 XP_006231668.1 5 - 6 694 90
GenBank mRNA AF003008.1 - 5 - 6 517 90
BC081824.1 - 5 - 6 564 90

View Full Product Details