Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd2
Entrez Gene ID
309374
Gene Name
ankyrin repeat domain 2 (stretch responsive muscle)
Gene Aliases
-
Chromosome Location
Chr.1: 261281859 - 261291307 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107589.1 NP_001101059.1 8 - 9 849 140
GenBank mRNA FQ216123.1 - 8 - 9 859 140
FQ216298.1 - 8 - 9 859 140
FQ216805.1 - 8 - 9 859 140

View Full Product Details