The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107599.1 | NP_001101069.1 | 4 - 5 | 2454 | 83 |
XM_008760429.1 | XP_008758651.1 | 4 - 5 | 2514 | 83 | |
XM_008760430.1 | XP_008758652.1 | 4 - 5 | 514 | 83 | |
XM_008760431.1 | XP_008758653.1 | 4 - 5 | 632 | 83 |