The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108526.1 | NP_001101996.1 | 13 - 14 | 2047 | 73 |
XM_006231528.2 | XP_006231590.1 | 14 - 15 | 2159 | 73 |