The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053735.1 | NP_446187.1 | 8 - 9 | 1329 | 98 |
GenBank mRNA | AY029199.1 | - | 8 - 9 | 1329 | 98 |