Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vwa2
Entrez Gene ID
307988
Gene Name
von Willebrand factor A domain containing 2
Gene Aliases
RGD1562000, Vwa2-ps1
Chromosome Location
Chr.1: 277689737 - 277730406 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003749146.3 XP_003749194.3 11 - 12 1732 83

View Full Product Details