Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sertad4
Entrez Gene ID
360899
Gene Name
SERTA domain containing 4
Gene Aliases
RGD1565408
Chromosome Location
Chr.13: 111574545 - 111585116 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108351.1 NP_001101821.1 2 - 3 363 92
XM_008769863.1 XP_008768085.1 1 - 2 4269 92
XM_008769864.1 XP_008768086.1 2 - 3 3739 92

View Full Product Details