The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126093.2 | NP_001119565.1 | 2 - 3 | 252 | 63 |
XM_008769760.1 | XP_008767982.1 | 2 - 3 | 1093 | 63 | |
GenBank mRNA | BC086330.1 | - | 2 - 3 | 252 | 63 |
BC162039.1 | - | 2 - 3 | 94 | 63 |