The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105986.1 | NP_001099456.1 | 4 - 5 | 538 | 111 |
GenBank mRNA | FQ211186.1 | - | 4 - 5 | 493 | 111 |
FQ216980.1 | - | 1 - 2 | 121 | 111 |