Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rrp15
Entrez Gene ID
360895
Gene Name
ribosomal RNA processing 15 homolog (S. cerevisiae)
Gene Aliases
RGD1306106
Chromosome Location
Chr.13: 105155824 - 105178907 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009702.1 NP_001009702.1 1 - 2 150 132
GenBank mRNA BC087649.1 - 1 - 2 150 132
FQ215627.1 - 1 - 2 149 132
FQ219085.1 - 1 - 2 171 131
FQ225526.1 - 1 - 2 174 132

View Full Product Details