Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rrp15
Entrez Gene ID
360895
Gene Name
ribosomal RNA processing 15 homolog (S. cerevisiae)
Gene Aliases
RGD1306106
Chromosome Location
Chr.13: 105155824 - 105178907 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009702.1 NP_001009702.1 2 - 3 402 76
GenBank mRNA BC087649.1 - 2 - 3 402 76
FQ215627.1 - 2 - 3 401 76
FQ219085.1 - 2 - 3 423 76
FQ225526.1 - 2 - 3 426 76

View Full Product Details